missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Difluoromethoxy)aniline, 97%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals B2034909

Catalog No. AAB2034909


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Difluoromethoxy)aniline
1.286
>110°C (230°F)
C7H7F2NO
MFCD00085005
UN2810
4-difluoromethoxy aniline, 4-difluoromethoxy-phenylamine, p-difluoromethoxyaniline, 4-amino-alpha,alpha-difluoroanisole, 4-difluoromethoxy benzenamine, benzenamine, 4-difluoromethoxy, 4-difluoromethoxy phenylamine, 4-difluormethoxy anilin, pubchem8467
C1=CC(=CC=C1N)OC(F)F
159.136
159.14
22236-10-8
231°C
Characteristic
1.5052
10 g
2804462
NDEZTSHWEPQVBX-UHFFFAOYSA-N
4-(difluoromethoxy)aniline
737363
97%

Chemical Identifiers

22236-10-8
159.136
NDEZTSHWEPQVBX-UHFFFAOYSA-N
737363
C1=CC(=CC=C1N)OC(F)F
C7H7F2NO
MFCD00085005
4-difluoromethoxy aniline, 4-difluoromethoxy-phenylamine, p-difluoromethoxyaniline, 4-amino-alpha,alpha-difluoroanisole, 4-difluoromethoxy benzenamine, benzenamine, 4-difluoromethoxy, 4-difluoromethoxy phenylamine, 4-difluormethoxy anilin, pubchem8467
4-(difluoromethoxy)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only