Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Difluoromethoxy)-3-methoxybenzaldehyde, 97%

$221.72 - $221.72

Chemical Identifiers

Chemical Name or Material 4-(Difluoromethoxy)-3-methoxybenzaldehyde
CAS 162401-70-9
Molecular Formula C9H8F2O3
Formula Weight 202.15g/mol
MDL Number MFCD03412217
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3284906

 
alfa aesar™
H32849-06
5g Each for $221.72
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 4-(Difluoromethoxy)-3-methoxybenzaldehyde
CAS 162401-70-9
Assay 97%
Molecular Formula C9H8F2O3
Formula Weight 202.15g/mol
MDL Number MFCD03412217
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.