Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Difluoromethoxy-2-fluorobenzoic acid, 97%

$80.00 - $266.70

Chemical Identifiers

Chemical Name or Material 4-Difluoromethoxy-2-fluorobenzoic acid
CAS 1214347-60-0
Molecular Formula C8H5F3O3
Formula Weight 206.12
MDL Number MFCD09832367
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3260003

  SDS
alfa aesar™
H3260003
1g Each for $266.70

AAH32600MD

  SDS
alfa aesar™
H32600MD
250mg Each for $80.00
Description & Specifications

Specifications

Formula Weight 206.12
Melting Point 135° to 138°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD09832367
Assay 97%
Chemical Name or Material 4-Difluoromethoxy-2-fluorobenzoic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
CAS 1214347-60-0
Molecular Formula C8H5F3O3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.