Learn More
4-Diethylaminosalicylaldehyde, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1165914
Description
4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.
Solubility
Insoluble in water.
Notes
It is sensitive to air. Store at room temperature. Incompatible with strong bases and strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
4-Diethylaminosalicylaldehyde | |
60°C to 64°C | |
MFCD00003326 | |
2212234 | |
4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde | |
XFVZSRRZZNLWBW-UHFFFAOYSA-N | |
4-(diethylamino)-2-hydroxybenzaldehyde | |
87293 | |
99% |
17754-90-4 | |
C11H15NO2 | |
25 g | |
Air Sensitive | |
Insoluble in water. | |
CCN(CC)C1=CC(=C(C=C1)C=O)O | |
193.246 | |
193.25 |
Chemical Identifiers
17754-90-4 | |
193.246 | |
XFVZSRRZZNLWBW-UHFFFAOYSA-N | |
87293 | |
CCN(CC)C1=CC(=C(C=C1)C=O)O |
C11H15NO2 | |
MFCD00003326 | |
4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde | |
4-(diethylamino)-2-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 241-745-3
RTECSNumber : CU5615150
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only