missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Diethylaminophenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1242006

 View more versions of this product

Catalog No. AAL1242006



Specifications

Specifications

4-Diethylaminophenyl isothiocyanate
147°C to 148°C (1.2mmHg)
5g
C11H14N2S
UN2923
4-diethylaminophenyl isothiocyanate, 4-diethylaminophenylisothiocyanate, 4-diethylamino benzenisothiocyanate, n,n-diethyl-4-isothiocyanato-aniline, #, benzenamine,n,n-diethyl-4-isothiocyanato, benzenamine, n,n-diethyl-4-isothiocyanato
CCN(CC)C1=CC=C(C=C1)N=C=S
206.307
206.31
>110°C (230°F)
30°C to 33°C
84381-54-4
MFCD00041094
3268361
PGWXUTHWTCRUAM-UHFFFAOYSA-N
N,N-diethyl-4-isothiocyanatoaniline
145027
97%

Chemical Identifiers

84381-54-4
206.307
PGWXUTHWTCRUAM-UHFFFAOYSA-N
145027
CCN(CC)C1=CC=C(C=C1)N=C=S
C11H14N2S
MFCD00041094
4-diethylaminophenyl isothiocyanate, 4-diethylaminophenylisothiocyanate, 4-diethylamino benzenisothiocyanate, n,n-diethyl-4-isothiocyanato-aniline, #, benzenamine,n,n-diethyl-4-isothiocyanato, benzenamine, n,n-diethyl-4-isothiocyanato
N,N-diethyl-4-isothiocyanatoaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents