missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Diethylaminobenzonitrile, 97%

$73.57 - $242.11

Chemical Identifiers

CAS 2873-90-7
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD00019754
InChI Key KMLGFOAKCYHXCQ-UHFFFAOYSA-N
Synonym 4-diethylamino benzonitrile, p-diethylaminobenzonitrile, benzonitrile, 4-diethylamino, 4-diethylamino benzenecarbonitrile, acmc-1cdnc, maybridge3_004418
PubChem CID 76134
IUPAC Name 4-(diethylamino)benzonitrile
SMILES CCN(CC)C1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0190506 View Documents Alfa Aesar™
L0190506
5g Each for $73.57
 
AAL0190514 View Documents Alfa Aesar™
L0190514
25g Each for $242.11
 
Specifications

Chemical Identifiers

2873-90-7
174.247
KMLGFOAKCYHXCQ-UHFFFAOYSA-N
76134
CCN(CC)C1=CC=C(C=C1)C#N
C11H14N2
MFCD00019754
4-diethylamino benzonitrile, p-diethylaminobenzonitrile, benzonitrile, 4-diethylamino, 4-diethylamino benzenecarbonitrile, acmc-1cdnc, maybridge3_004418
4-(diethylamino)benzonitrile

Specifications

66°C to 67°C
2873-90-7
MFCD00019754
973913
KMLGFOAKCYHXCQ-UHFFFAOYSA-N
4-(diethylamino)benzonitrile
76134
97%
5g
C11H14N2
UN3439
4-diethylamino benzonitrile, p-diethylaminobenzonitrile, benzonitrile, 4-diethylamino, 4-diethylamino benzenecarbonitrile, acmc-1cdnc, maybridge3_004418
CCN(CC)C1=CC=C(C=C1)C#N
174.247
174.25
4-Diethylaminobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.