Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Diethylaminobenzoic acid, 98%

$69.46 - $189.85

Chemical Identifiers

Chemical Name or Material 4-Diethylaminobenzoic acid
CAS 5429-28-7
Molecular Formula C11H15NO2
Formula Weight 193.25
MDL Number MFCD00002538
Beilstein 2211601
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1290314

  SDS
alfa aesar™
A1290314
25g Each for $69.46

AAA1290322

  SDS
alfa aesar™
A1290322
100g Each for $189.85
Description & Specifications

Specifications

Formula Weight 193.25
Chemical Name or Material 4-Diethylaminobenzoic acid
TSCA TSCA
Assay 98%
Molecular Formula C11H15NO2
MDL Number MFCD00002538
CAS 5429-28-7
Beilstein 2211601
Melting Point 192° to 195°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g

Applications
4-Diethylaminobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in hot methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.