missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Diethylaminobenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ A1182522

 View more versions of this product

Catalog No. AAA1182522


 

Description

Description

Applications
4-Diethylaminobenzaldehyde is used in the preparation of 5-(4-diethylamino-benzylidene)-selenazolidine-2,4-dione by using aqueous methylamine as a reagent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

4-Diethylaminobenzaldehyde
120-21-8
C11H15NO
511102
39°C to 41°C
MNFZZNNFORDXSV-UHFFFAOYSA-N
4-(diethylamino)benzaldehyde
67114
177.25
100g
97%
MFCD00003382
173°C to 175°C (7mmHg)
165°C (329°F)
CCN(CC)C1=CC=C(C=C1)C=O
177.247
CHEBI:86194

Chemical Identifiers

120-21-8
177.247
MNFZZNNFORDXSV-UHFFFAOYSA-N
CHEBI:86194
C11H15NO
MFCD00003382
67114
Safety and Handling

Safety and Handling

GHS H Statement
H311-H315-H319-H335
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : CU5612850

TSCA : TSCA

SDS Documents