Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Diethylaminoacetophenone, 99%

$65.73 - $213.96

Chemical Identifiers

Chemical Name or Material 4'-Diethylaminoacetophenone
CAS 5520-66-1
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD00043651
Beilstein 2093988
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1333103

  SDS
alfa aesar™
L1333103
1g Each for $65.73

AAL1333106

  SDS
alfa aesar™
L1333106
5g Each for $213.96
Description & Specifications

Specifications

Formula Weight 191.27
Chemical Name or Material 4'-Diethylaminoacetophenone
Assay 99%
Molecular Formula C12H17NO
MDL Number MFCD00043651
CAS 5520-66-1
Beilstein 2093988
Melting Point 47° to 49°C
Quantity 1g