Please login to your online account to display your discounted pricing

4-(Diethylamino)benzoic acid, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC217730010

 View more versions of this product

Catalog No. AC217730010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 193.24
Quantity 1g
Color Light green
EINECS Number 226-582-8
TSCA TSCA
Melting Point 192° to 195°C
Beilstein 14, 429
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-(Diethylamino)benzoic acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C11H15NO2
Linear Formula (C2H5)2NC6H4CO2H
MDL Number MFCD00002538
CAS 5429-28-7

Specifications

Chemical Name or Material 4-(Diethylamino)benzoic acid
Name Note 99%
CAS 5429-28-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H15NO2
Linear Formula (C2H5)2NC6H4CO2H
Formula Weight 193.24
MDL Number MFCD00002538
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents