Please login to your online account to display your discounted pricing

4-(Diethylamino)benzoic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Diethylamino)benzoic acid
Name Note 99%
CAS 5429-28-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H15NO2
Linear Formula (C2H5)2NC6H4CO2H
Formula Weight 193.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC217730010

  SDS
thermo fisher scientific
AC217730010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC217731000

  SDS
thermo fisher scientific
217731000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 193.24
Quantity 1g
Color Light green
EINECS Number 226-582-8
TSCA TSCA
Melting Point 192° to 195°C
Beilstein 14, 429
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-(Diethylamino)benzoic acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C11H15NO2
Linear Formula (C2H5)2NC6H4CO2H
MDL Number MFCD00002538
CAS 5429-28-7
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.