missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, Laser Grade 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 407075000

 View more versions of this product

Catalog No. AC407075000


 

Specifications Description & Specifications

Specifications

≥96.0%
Extra Pure
500mg
15, 2841
96.0
480nm (in DMSO)
37700L/(mole.cm) (lambda max.)
Authentic
644nm (in DMSO)
99%
96+%
Powder or Crystalline Needles
303.35
Dark red
100.0
(for use as a laser dye) passes
661nm (in DMSO)
DCM
4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran
C19H17N3O
MFCD00051341
51325-91-8

Chemical Identifiers

51325-91-8
100.0
C19H17N3O
MFCD00051341
4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, 99%
Extra Pure
96.0
96+%
303.35
DCM
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 257-137-6

RTECSNumber : OO3746100

TSCA : TSCA