missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Dibutylamino)benzaldehyde, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439770010
Specifications
4-(Dibutylamino)benzaldehyde | |
Authentic | |
148.0°C (1.0 Torr) | |
1g | |
97.5 | |
97.5% min. (GC) | |
4-dibutylamino benzaldehyde, 4-dibutylamino-benzaldehyde, 4-dibutylaminobenzaldehyd, 4-dibutylaminobenzoic aldehyde | |
CCCCN(CCCC)C1=CC=C(C=C1)C=O | |
233.35 | |
233.35 | |
98% |
113°C | |
Glass bottle | |
Yellow to Brown | |
90134-10-4 | |
100.0 | |
C15H23NO | |
VNWPLOWYHIDMEB-UHFFFAOYSA-N | |
4-(dibutylamino)benzaldehyde | |
605391 | |
Liquid |
Chemical Identifiers
90134-10-4 | |
233.35 | |
4-dibutylamino benzaldehyde, 4-dibutylamino-benzaldehyde, 4-dibutylaminobenzaldehyd, 4-dibutylaminobenzoic aldehyde | |
4-(dibutylamino)benzaldehyde |
C15H23NO | |
VNWPLOWYHIDMEB-UHFFFAOYSA-N | |
605391 | |
CCCCN(CCCC)C1=CC=C(C=C1)C=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
GHS Signal Word: Warning