missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Dibutylamino)benzaldehyde, 98%, ACROS Organics™

Chemical Identifiers

CAS 90134-10-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C15H23NO
Formula Weight 233.35
Chemical Name or Material 4-(Dibutylamino)benzaldehyde
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC439770010 SDS Thermo Fisher Scientific
AC439770010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439770050 SDS Thermo Fisher Scientific
AC439770050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

90134-10-4
100.0
C15H23NO
4-(Dibutylamino)benzaldehyde
97.5
98%
233.35
≥97.5%

Specifications

Liquid
1g
100.0
90134-10-4
C15H23NO
Glass bottle
≥97.5%
233.35
Yellow to brown
Authentic
97.5
98%
4-(Dibutylamino)benzaldehyde

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.