Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Dibenzylamino)benzeneboronic acid, 95%

$62.84 - $138.47

Chemical Identifiers

Chemical Name or Material 4-(Dibenzylamino)benzeneboronic acid
CAS 159191-44-3
Molecular Formula C20H20BNO2
Formula Weight 317.19
MDL Number MFCD06798225
Synonym 4-(Dibenzylamino)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2755003

  SDS
alfa aesar™
H2755003
1g Each for $138.47

AAH27550MD

  SDS
alfa aesar™
H27550MD
250mg Each for $62.84
Description & Specifications

Specifications

Formula Weight 317.19
MDL Number MFCD06798225
Quantity 1g
Melting Point 184° to 188°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Chemical Name or Material 4-(Dibenzylamino)benzeneboronic acid
CAS 159191-44-3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C20H20BNO2
Synonym 4-(Dibenzylamino)phenylboronic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.