Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Di-n-propylsulfamoyl)benzoyl chloride, 96%

Chemical Identifiers

Chemical Name or Material 4-(Di-n-propylsulfamoyl)benzoyl chloride
CAS 29171-72-0
Molecular Formula C13H18ClNO3S
Formula Weight 303.81
MDL Number MFCD00121817
UN Number UN3261
Synonym 4-(Di-n-propylaminosulfonyl)benzoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5070303

 
alfa aesar™
H5070303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50703MD

 
alfa aesar™
H50703MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 303.81
Chemical Name or Material 4-(Di-n-propylsulfamoyl)benzoyl chloride
Assay 96%
Molecular Formula C13H18ClNO3S
MDL Number MFCD00121817
CAS 29171-72-0
UN Number UN3261
Synonym 4-(Di-n-propylaminosulfonyl)benzoyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 59° to 61°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.