missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Di-n-propylsulfamoyl)acetanilide, 97%

Chemical Identifiers

CAS 294885-63-5
Molecular Formula C14H22N2O3S
Molecular Weight (g/mol) 298.401
MDL Number MFCD00577457
InChI Key RSKJZTBKLIORJO-UHFFFAOYSA-N
Synonym 4'-di-n-propylsulfamoyl acetanilide
PubChem CID 668753
IUPAC Name N-[4-(dipropylsulfamoyl)phenyl]acetamide
SMILES CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)NC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60353MD View Documents Alfa Aesar™
H60353MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6035303 View Documents Alfa Aesar™
H6035303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

294885-63-5
298.401
RSKJZTBKLIORJO-UHFFFAOYSA-N
668753
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)NC(=O)C
C14H22N2O3S
MFCD00577457
4'-di-n-propylsulfamoyl acetanilide
N-[4-(dipropylsulfamoyl)phenyl]acetamide

Specifications

250mg
C14H22N2O3S
4'-di-n-propylsulfamoyl acetanilide
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)NC(=O)C
298.401
298.4
4'-(Di-n-propylsulfamoyl)acetanilide
294885-63-5
MFCD00577457
RSKJZTBKLIORJO-UHFFFAOYSA-N
N-[4-(dipropylsulfamoyl)phenyl]acetamide
668753
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning