Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Decyne, 98%

Chemical Identifiers

Chemical Name or Material 4-Decyne
CAS 2384-86-3
Molecular Formula CH3(CH2)4C≡CCH2CH2CH3
Formula Weight 138.26
MDL Number MFCD00039978
UN Number UN3295
Beilstein 1698308
Synonym n-Pentyl n-propyl acetylene; n-Amyl n-propyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2059706

 
alfa aesar™
B2059706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2059714

 
alfa aesar™
B2059714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.26
Chemical Name or Material 4-Decyne
Assay 98%
Molecular Formula CH3(CH2)4C≡CCH2CH2CH3
MDL Number MFCD00039978
CAS 2384-86-3
UN Number UN3295
Beilstein 1698308
Synonym n-Pentyl n-propyl acetylene; n-Amyl n-propyl acetylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 75° to 76°C (20mmHg)
Flash Point 52°C (125°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Quantity 5g

Applications
4-Decyne is used in acid zinc plating.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.