Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Decanol, 97+%

$62.84 - $631.34

Chemical Identifiers

Chemical Name or Material 4-Decanol
CAS 2051-31-2
Molecular Formula C10H22O
Formula Weight 158.29
MDL Number MFCD00039627
Beilstein 1735252
Synonym n-Hexyl n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2086606

  SDS
alfa aesar™
B2086606
5g Each for $62.84

AAB2086614

  SDS
alfa aesar™
B2086614
25g Each for $206.43

AAB2086622

  SDS
alfa aesar™
B2086622
100g Each for $631.34
Description & Specifications

Specifications

Formula Weight 158.29
Beilstein 1735252
Flash Point 82°C (179°F)
Synonym n-Hexyl n-propyl carbinol
Assay ≥97%
MDL Number MFCD00039627
Chemical Name or Material 4-Decanol
Molecular Formula C10H22O
CAS 2051-31-2
Quantity 5g
Boiling Point 210° to 211°C
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.