missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Cyclopropylsulfamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53170MD

 View more versions of this product

Catalog No. AAH53170MD



Description

Description

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

4-(Cyclopropylsulfamoyl)benzeneboronic acid
250mg
C9H12BNO4S
n-cyclopropyl 4-boronobenzenesulfonamide, 4-cyclopropylsulfamoyl phenylboronic acid, 4-n-cyclopropylsulfamoyl phenyl boronic acid, 4-n-cyclopropylsulfamoyl phenylboronic acid, 4-cyclopropylsulfamoyl phenyl boronic acid, 4-borono-n-cyclopropylbenzenesulphonamide, 4-n-cyclopropylsulphonamido benzeneboronic acid, boronic acid,b-4-cyclopropylamino sulfonyl phenyl, acmc-209qgb, 4-n-cyclopropylsulfamoyl phenyl boronicacid
B(C1=CC=C(C=C1)S(=O)(=O)NC2CC2)(O)O
241.068
241.07
142°C to 144°C
871329-67-8
MFCD07363755
FUEGEHDCXRTJPN-UHFFFAOYSA-N
[4-(cyclopropylsulfamoyl)phenyl]boronic acid
44119553
98%

Chemical Identifiers

871329-67-8
241.068
FUEGEHDCXRTJPN-UHFFFAOYSA-N
44119553
B(C1=CC=C(C=C1)S(=O)(=O)NC2CC2)(O)O
C9H12BNO4S
MFCD07363755
n-cyclopropyl 4-boronobenzenesulfonamide, 4-cyclopropylsulfamoyl phenylboronic acid, 4-n-cyclopropylsulfamoyl phenyl boronic acid, 4-n-cyclopropylsulfamoyl phenylboronic acid, 4-cyclopropylsulfamoyl phenyl boronic acid, 4-borono-n-cyclopropylbenzenesulphonamide, 4-n-cyclopropylsulphonamido benzeneboronic acid, boronic acid,b-4-cyclopropylamino sulfonyl phenyl, acmc-209qgb, 4-n-cyclopropylsulfamoyl phenyl boronicacid
[4-(cyclopropylsulfamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents