Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyclopropyl-4-oxobutyric acid, 95%

$218.58 - $218.58

Chemical Identifiers

Chemical Name or Material 4-Cyclopropyl-4-oxobutyric acid
CAS 53712-75-7
Molecular Formula C7H10O3
Formula Weight 142.15
MDL Number MFCD01320151
Synonym 3-Cyclopropanoylpropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5107803

 
alfa aesar™
H5107803
1g Each for $218.58
Description & Specifications

Specifications

Formula Weight 142.15
Chemical Name or Material 4-Cyclopropyl-4-oxobutyric acid
Assay 95%
Molecular Formula C7H10O3
MDL Number MFCD01320151
Synonym 3-Cyclopropanoylpropionic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 53712-75-7
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.