missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5449003

 View more versions of this product

Catalog No. AAH5449003



Specifications

Specifications

4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid
1221823-27-3
MFCD20265279
JDFPAMLPRYKLRV-UHFFFAOYSA-N
[4-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
57655465
97%
1g
C13H20BNO4S
4-cyclohexylmethylsulfamoyl benzeneboronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NCC2CCCCC2)(O)O
297.176
297.18

Chemical Identifiers

1221823-27-3
297.176
JDFPAMLPRYKLRV-UHFFFAOYSA-N
57655465
B(C1=CC=C(C=C1)S(=O)(=O)NCC2CCCCC2)(O)O
C13H20BNO4S
MFCD20265279
4-cyclohexylmethylsulfamoyl benzeneboronic acid
[4-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents