missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester

$42.96 - $663.43

Chemical Identifiers

CAS 1218790-50-1
Molecular Formula C19H28BNO2
Formula Weight 313.24
MDL Number MFCD11113042
Synonym 4-Cyclohexyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
Chemical Name or Material 4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5112703 SDS Alfa Aesar™
H5112703
1g Each for $42.96
AAH5112706 SDS Alfa Aesar™
H5112706
5g Each for $166.24
AAH5112714 SDS Alfa Aesar™
H5112714
25g Each for $663.43
Specifications Description & Specifications

Chemical Identifiers

1218790-50-1
313.24
4-Cyclohexyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
C19H28BNO2
MFCD11113042
4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester

Specifications

1g
C19H28BNO2
MFCD11113042
4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester
1218790-50-1
313.24
4-Cyclohexyliminomethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene