missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyclohexylbenzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438000050

 View more versions of this product

Catalog No. AC438000050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Cyclohexylbenzenesulfonyl chloride
White
5g
96.0
96% min. as diethylsulfonamide (HPLC)
4-cyclohexyl-1-benzenesulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexylphenylsulfonyl chloride, acmc-1awu9, benzenesulfonyl chloride, 4-cyclohexyl, chloro 4-cyclohexylphenyl sulfone
C1CCC(CC1)C2=CC=C(C=C2)S(=O)(=O)Cl
258.77
258.77
97%
Authentic
54.2°C to 55.7°C
56354-57-5
100.0
C12H15ClO2S
CREMYEDHKUWVTB-UHFFFAOYSA-N
4-cyclohexylbenzenesulfonyl chloride
4339442
Crystalline Powder

Chemical Identifiers

56354-57-5
258.77
4-cyclohexyl-1-benzenesulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexylphenylsulfonyl chloride, acmc-1awu9, benzenesulfonyl chloride, 4-cyclohexyl, chloro 4-cyclohexylphenyl sulfone
4-cyclohexylbenzenesulfonyl chloride
C12H15ClO2S
CREMYEDHKUWVTB-UHFFFAOYSA-N
4339442
C1CCC(CC1)C2=CC=C(C=C2)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Danger

SDS Documents