missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Cyclohexylbenzenesulfonyl chloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438000010
Specifications
4-Cyclohexylbenzenesulfonyl chloride | |
Glass Bottle | |
54.2°C to 55.7°C | |
56354-57-5 | |
100.0 | |
C12H15ClO2S | |
CREMYEDHKUWVTB-UHFFFAOYSA-N | |
4-cyclohexylbenzenesulfonyl chloride | |
4339442 | |
Crystalline Powder |
Authentic | |
White | |
1g | |
96.0 | |
96% min. as diethylsulfonamide (HPLC) | |
4-cyclohexyl-1-benzenesulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexylphenylsulfonyl chloride, acmc-1awu9, benzenesulfonyl chloride, 4-cyclohexyl, chloro 4-cyclohexylphenyl sulfone | |
C1CCC(CC1)C2=CC=C(C=C2)S(=O)(=O)Cl | |
258.77 | |
258.77 | |
97% |
Chemical Identifiers
56354-57-5 | |
258.77 | |
4-cyclohexyl-1-benzenesulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexylphenylsulfonyl chloride, acmc-1awu9, benzenesulfonyl chloride, 4-cyclohexyl, chloro 4-cyclohexylphenyl sulfone | |
4-cyclohexylbenzenesulfonyl chloride |
C12H15ClO2S | |
CREMYEDHKUWVTB-UHFFFAOYSA-N | |
4339442 | |
C1CCC(CC1)C2=CC=C(C=C2)S(=O)(=O)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
GHS Signal Word: Danger