missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyclohexylbenzenesulfonyl chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 56354-57-5
Molecular Formula C12H15ClO2S
Molecular Weight (g/mol) 258.77
InChI Key CREMYEDHKUWVTB-UHFFFAOYSA-N
Synonym 4-cyclohexyl-1-benzenesulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexylphenylsulfonyl chloride, acmc-1awu9, benzenesulfonyl chloride, 4-cyclohexyl, chloro 4-cyclohexylphenyl sulfone
PubChem CID 4339442
IUPAC Name 4-cyclohexylbenzenesulfonyl chloride
SMILES C1CCC(CC1)C2=CC=C(C=C2)S(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC438000010
SDS View Documents
Thermo Fisher Scientific
AC438000010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC438000050
SDS View Documents
Thermo Fisher Scientific
AC438000050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

56354-57-5
258.77
4-cyclohexyl-1-benzenesulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexylphenylsulfonyl chloride, acmc-1awu9, benzenesulfonyl chloride, 4-cyclohexyl, chloro 4-cyclohexylphenyl sulfone
4-cyclohexylbenzenesulfonyl chloride
C12H15ClO2S
CREMYEDHKUWVTB-UHFFFAOYSA-N
4339442
C1CCC(CC1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Authentic
White
1g
96.0
96% min. as diethylsulfonamide (HPLC)
4-cyclohexyl-1-benzenesulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexylphenylsulfonyl chloride, acmc-1awu9, benzenesulfonyl chloride, 4-cyclohexyl, chloro 4-cyclohexylphenyl sulfone
C1CCC(CC1)C2=CC=C(C=C2)S(=O)(=O)Cl
258.77
258.77
97%
Glass Bottle
54.2°C to 55.7°C
56354-57-5
100.0
C12H15ClO2S
CREMYEDHKUWVTB-UHFFFAOYSA-N
4-cyclohexylbenzenesulfonyl chloride
4339442
Crystalline Powder
4-Cyclohexylbenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Danger

SDS