Learn More
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 292910025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Cyanostyrene | |
Colorless to Yellow | |
Authentic | |
Glass bottle | |
1.5765 to 1.5785 | |
2.5 g | |
Solubility in water: insoluble | |
C=CC1=CC=C(C=C1)C#N | |
129.16 | |
76967 | |
95% |
3435-51-6 | |
56.0°C to 58.0°C (10.0 mmHg) | |
94% min. (GC) | |
C9H7N | |
MFCD00080445 | |
4-cyanostyrene, 4-vinylbenzonitrile, benzonitrile, 4-ethenyl, p-cyanostyrene, 4-vinyl-benzonitrile, benzonitrile, p-vinyl, p-vinylbenzonitrile, acmc-20amk5, benzonitrile,4-ethenyl, 4-vinylbenzenecarbonitrile | |
SNTUCKQYWGHZPK-UHFFFAOYSA-N | |
4-ethenylbenzonitrile | |
0.05% tert.butylcatechol | |
129.16 | |
Liquid |
Chemical Identifiers
3435-51-6 | |
129.16 | |
SNTUCKQYWGHZPK-UHFFFAOYSA-N | |
4-ethenylbenzonitrile |
C9H7N | |
MFCD00080445 | |
76967 | |
C=CC1=CC=C(C=C1)C#N |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO – Research Use Only