missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Cyanostyrene, 95%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC292910100
Specifications
4-Cyanostyrene | |
Glass bottle | |
0.05% tert.butylcatechol | |
Colorless to Yellow | |
3435-51-6 | |
C9H7N | |
Solubility in water: insoluble | |
C=CC1=CC=C(C=C1)C#N | |
129.16 | |
129.16 | |
95% |
Authentic | |
1.5765 to 1.5785 | |
56.0°C to 58.0°C (10.0mmHg) | |
10g | |
94% min. (GC) | |
4-cyanostyrene, 4-vinyl-benzonitrile, 4-vinylbenzenecarbonitrile, 4-vinylbenzonitrile, acmc-20amk5, benzonitrile, 4-ethenyl, benzonitrile, p-vinyl, benzonitrile,4-ethenyl, p-cyanostyrene, p-vinylbenzonitrile | |
SNTUCKQYWGHZPK-UHFFFAOYSA-N | |
4-ethenylbenzonitrile | |
76967 | |
Liquid |
Chemical Identifiers
3435-51-6 | |
129.16 | |
4-cyanostyrene, 4-vinyl-benzonitrile, 4-vinylbenzenecarbonitrile, 4-vinylbenzonitrile, acmc-20amk5, benzonitrile, 4-ethenyl, benzonitrile, p-vinyl, benzonitrile,4-ethenyl, p-cyanostyrene, p-vinylbenzonitrile | |
4-ethenylbenzonitrile |
C9H7N | |
SNTUCKQYWGHZPK-UHFFFAOYSA-N | |
76967 | |
C=CC1=CC=C(C=C1)C#N |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning