Learn More
4-Cyanostyrene, 97%, stab. with 0.05% 4-tert-butyl catechol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2439514
Description
4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.
Solubility
Immiscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with polymerizing initiators, polymerization catalysts and strong oxidizing agents.
Specifications
4-Cyanostyrene | |
3435-51-6 | |
C9H7N | |
MFCD00080445 | |
UN3276 | |
Immiscible with water. | |
C=CC1=CC=C(C=C1)C#N | |
129.16 | |
129.16 |
stabilized with 0.05% 4-tert-butyl catechol | |
92°C to 93°C (3 mmHg) | |
1.5775 | |
25 g | |
4-cyanostyrene, 4-vinylbenzonitrile, benzonitrile, 4-ethenyl, p-cyanostyrene, 4-vinyl-benzonitrile, benzonitrile, p-vinyl, p-vinylbenzonitrile, acmc-20amk5, benzonitrile,4-ethenyl, 4-vinylbenzenecarbonitrile | |
SNTUCKQYWGHZPK-UHFFFAOYSA-N | |
4-ethenylbenzonitrile | |
76967 | |
97% |
Chemical Identifiers
3435-51-6 | |
129.16 | |
SNTUCKQYWGHZPK-UHFFFAOYSA-N | |
4-ethenylbenzonitrile |
C9H7N | |
MFCD00080445 | |
76967 | |
C=CC1=CC=C(C=C1)C#N |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only