Please login to your online account to display your discounted pricing

4-Cyanopyridine 98%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  110871000

 View more versions of this product

Catalog No. AC110871000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals or Powder
Assay ≥97.5% (GC)
Formula Weight 104.11
Quantity 100g
Color White to off-white
TSCA TSCA
EINECS Number 202-856-2
Melting Point 76° to 80°C
CAS Max % 100.0
Assay Percent Range 98%
Water 0.5% max. (K.F.)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Health Hazard 1 Warning
CAS 100-48-1
Name Note 98%
CAS Min % 97.5
Molecular Formula C6H4N2
MDL Number MFCD00006417
Chemical Name or Material 4-Cyanopyridine
Linear Formula (C5H4N)CN
Packaging Plastic bottle
Synonym Isonicotinonitrile
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Cyanopyridine
Name Note 98%
CAS 100-48-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H4N2
Linear Formula (C5H4N)CN
Formula Weight 104.11
MDL Number MFCD00006417
Synonym Isonicotinonitrile
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents