Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanopyridine-3-boronic acid pinacol ester

$256.15 - $580.11

Chemical Identifiers

Chemical Name or Material 4-Cyanopyridine-3-boronic acid pinacol ester
CAS 878194-91-3
Molecular Formula C12H15BN2O2
Formula Weight 230.07
MDL Number MFCD08458478
Synonym 4-Cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27216MD

  SDS
alfa aesar™
H27216MD
250mg Each for $256.15

AAH2721603

  SDS
alfa aesar™
H2721603
1g Each for $580.11
Description & Specifications

Specifications

Formula Weight 230.07
Chemical Name or Material 4-Cyanopyridine-3-boronic acid pinacol ester
Molecular Formula C12H15BN2O2
MDL Number MFCD08458478
CAS 878194-91-3
Synonym 4-Cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 84° to 88°C
Quantity 250mg
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.