missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanophenylhydrazine hydrochloride, 97%

$982.05 - $982.05

Chemical Identifiers

CAS 2863-98-1
Molecular Formula C7H7N3·HCl
Formula Weight 169.61
MDL Number MFCD00673994
Beilstein 3565486
Synonym 4-Hydrazinobenzonitrile hydrochloride
Chemical Name or Material 4-Cyanophenylhydrazine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2215722 SDS Alfa Aesar™
B2215722
100g Each for $982.05
AAB2215706 SDS Alfa Aesar™
B2215706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAB2215714 SDS Alfa Aesar™
B2215714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Description & Specifications

Chemical Identifiers

2863-98-1
169.61
3565486
4-Cyanophenylhydrazine hydrochloride
C7H7N3·HCl
MFCD00673994
4-Hydrazinobenzonitrile hydrochloride

Specifications

≈243°C (dec.)
2863-98-1
169.61
3565486
4-Cyanophenylhydrazine hydrochloride
100g
C7H7N3·HCl
MFCD00673994
4-Hydrazinobenzonitrile hydrochloride
97%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger