Learn More
4-Cyanophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2215714
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications
4-Cyanophenylhydrazine hydrochloride | |
∼243°C (decomposition) | |
MFCD00673994 | |
3565486 | |
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j | |
DZUUSHCOMPROCJ-UHFFFAOYSA-N | |
4-hydrazinylbenzonitrile;hydrochloride | |
16212962 | |
97% |
2863-98-1 | |
C7H7N3 | |
25 g | |
Moisture sensitive | |
Soluble in methanol. | |
NNC1=CC=C(C=C1)C#N | |
133.15 | |
169.61 |
Chemical Identifiers
2863-98-1 | |
133.15 | |
DZUUSHCOMPROCJ-UHFFFAOYSA-N | |
16212962 | |
NNC1=CC=C(C=C1)C#N |
C7H7N3 | |
MFCD00673994 | |
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j | |
4-hydrazinylbenzonitrile;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only