missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanophenylacetonitrile, 97%

Manufacturer:  Alfa Aesar™ H5411206

 View more versions of this product

Catalog No. AAH5411206



Specifications

Specifications

4-Cyanophenylacetonitrile
180°C to 181°C
876-31-3
MFCD03839848
QILKKAFYAFEWGU-UHFFFAOYSA-N
4-(cyanomethyl)benzonitrile
136686
97%
71°C (160°F)
5g
C9H6N2
.alpha.-cyano-p-tolunitrile
C1=CC(=CC=C1CC#N)C#N
142.161
142.16

Chemical Identifiers

876-31-3
142.161
QILKKAFYAFEWGU-UHFFFAOYSA-N
136686
C1=CC(=CC=C1CC#N)C#N
C9H6N2
MFCD03839848
.alpha.-cyano-p-tolunitrile
4-(cyanomethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents