Learn More
4-Cyanophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1017306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Cyanophenyl isothiocyanate | |
118°C to 122°C | |
C8H4N2S | |
5 g | |
2802621 | |
4-cyanophenyl isothiocyanate, benzonitrile, 4-isothiocyanato, p-cyanophenyl isothiocyanate, 4-cyanophenylisothiocyanate, 4-cyano-phenyl-isothiocyanate, 4-isothiocyanatobenzenenitrile, 4-isothiocyanato-benzonitrile, benzonitrile,4-isothiocyanato, isothiocyanic acid, p-cyanophenyl ester, benzenenitrile, 4-isothiocyanato | |
C1=CC(=CC=C1C#N)N=C=S | |
160.194 | |
160.2 |
2719-32-6 | |
Irritating | |
MFCD00041085 | |
UN2923 | |
Moisture sensitive | |
DZFKAXLNKZXNHD-UHFFFAOYSA-N | |
4-isothiocyanatobenzonitrile | |
17626 | |
98% |
Chemical Identifiers
2719-32-6 | |
160.194 | |
DZFKAXLNKZXNHD-UHFFFAOYSA-N | |
17626 | |
C1=CC(=CC=C1C#N)N=C=S |
C8H4N2S | |
MFCD00041085 | |
4-cyanophenyl isothiocyanate, benzonitrile, 4-isothiocyanato, p-cyanophenyl isothiocyanate, 4-cyanophenylisothiocyanate, 4-cyano-phenyl-isothiocyanate, 4-isothiocyanatobenzenenitrile, 4-isothiocyanato-benzonitrile, benzonitrile,4-isothiocyanato, isothiocyanic acid, p-cyanophenyl ester, benzenenitrile, 4-isothiocyanato | |
4-isothiocyanatobenzonitrile |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 220-323-2
RTECSNumber : NX8479500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only