missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanophenyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1017314

 View more versions of this product

Catalog No. AAL1017314



Specifications

Specifications

4-Cyanophenyl isothiocyanate
118°C to 122°C
2719-32-6
MFCD00041085
2802621
DZFKAXLNKZXNHD-UHFFFAOYSA-N
4-isothiocyanatobenzonitrile
17626
98%
Irritating
25g
C8H4N2S
UN2923
4-cyanophenyl isothiocyanate, benzonitrile, 4-isothiocyanato, p-cyanophenyl isothiocyanate, 4-cyanophenylisothiocyanate, 4-cyano-phenyl-isothiocyanate, 4-isothiocyanatobenzenenitrile, 4-isothiocyanato-benzonitrile, benzonitrile,4-isothiocyanato, isothiocyanic acid, p-cyanophenyl ester, benzenenitrile, 4-isothiocyanato
C1=CC(=CC=C1C#N)N=C=S
160.194
160.2

Chemical Identifiers

2719-32-6
160.194
DZFKAXLNKZXNHD-UHFFFAOYSA-N
17626
C1=CC(=CC=C1C#N)N=C=S
C8H4N2S
MFCD00041085
4-cyanophenyl isothiocyanate, benzonitrile, 4-isothiocyanato, p-cyanophenyl isothiocyanate, 4-cyanophenylisothiocyanate, 4-cyano-phenyl-isothiocyanate, 4-isothiocyanatobenzenenitrile, 4-isothiocyanato-benzonitrile, benzonitrile,4-isothiocyanato, isothiocyanic acid, p-cyanophenyl ester, benzenenitrile, 4-isothiocyanato
4-isothiocyanatobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : NX8479500

TSCA : TSCA

Documents