Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanophenyl 4-n-propylbenzoate, 99%

Chemical Identifiers

Chemical Name or Material 4-Cyanophenyl 4-n-propylbenzoate
CAS 56131-49-8
Molecular Formula C17H15NO2
Formula Weight 265.31
MDL Number MFCD03410282
UN Number UN3439
Synonym 4-n-Propylbenzoic acid 4-cyanophenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2227403

  SDS
alfa aesar™
B2227403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2227406

  SDS
alfa aesar™
B2227406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 265.31
Chemical Name or Material 4-Cyanophenyl 4-n-propylbenzoate
Quantity 1g
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
MDL Number MFCD03410282
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 4-n-Propylbenzoic acid 4-cyanophenyl ester
CAS 56131-49-8
Health Hazard 1 Danger
UN Number UN3439
Melting Point 101° to 103°C
Molecular Formula C17H15NO2
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.