missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Cyanomethyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC444740050

 View more versions of this product

Catalog No. AC444740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-(Cyanomethyl)phenylboronic acid pinacol ester
5g
97%
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC#N
243.113
243.12
77.0°C to 80.0°C
138500-86-4
C14H18BNO2
URWMFRYGXSHPRV-UHFFFAOYSA-N
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
3842922
97%

Chemical Identifiers

138500-86-4
243.113
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
C14H18BNO2
URWMFRYGXSHPRV-UHFFFAOYSA-N
3842922
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC#N
SDS Documents