Please login to your online account to display your discounted pricing

4-(Cyanomethyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  444740010

 View more versions of this product

Catalog No. AC444740010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material 4-(Cyanomethyl)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H18BNO2
Formula Weight 243.12g/mol

Specifications

Chemical Name or Material 4-(Cyanomethyl)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H18BNO2
Formula Weight 243.12g/mol