missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Cyanomethyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 138500-86-4
Molecular Formula C14H18BNO2
Molecular Weight (g/mol) 243.113
InChI Key URWMFRYGXSHPRV-UHFFFAOYSA-N
Synonym 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
PubChem CID 3842922
IUPAC Name 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444740050 SDS View Documents Acros Organics
AC444740050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

138500-86-4
243.113
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
C14H18BNO2
URWMFRYGXSHPRV-UHFFFAOYSA-N
3842922
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC#N

Specifications

77.0°C to 80.0°C
138500-86-4
C14H18BNO2
URWMFRYGXSHPRV-UHFFFAOYSA-N
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
3842922
97%
5g
97%
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC#N
243.113
243.12
4-(Cyanomethyl)phenylboronic acid pinacol ester