Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Cyanomethyl)benzeneboronic acid pinacol ester, 95%

$158.22 - $617.77

Chemical Identifiers

Chemical Name or Material 4-(Cyanomethyl)benzeneboronic acid pinacol ester
CAS 138500-86-4
Molecular Formula C14H18BNO2
Formula Weight 243.11
MDL Number MFCD02179459
Synonym 4-(Cyanomethyl)phenylboronic acid pinacol cyclic ester; 2-[(4-Cyanomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1995603

  SDS
alfa aesar™
L1995603
1g Each for $158.22

AAL1995606

  SDS
alfa aesar™
L1995606
5g Each for $617.77
Description & Specifications

Specifications

Formula Weight 243.11
CAS 138500-86-4
MDL Number MFCD02179459
Molecular Formula C14H18BNO2
Chemical Name or Material 4-(Cyanomethyl)benzeneboronic acid pinacol ester
Synonym 4-(Cyanomethyl)phenylboronic acid pinacol cyclic ester; 2-[(4-Cyanomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Health Hazard 1 Warning
Assay 95%
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Quantity 1g
Melting Point 74° to 78°C
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.