Learn More
4-(Cyanomethyl)benzeneboronic acid, 95%
CAS: 91983-26-5 | C8H8BNO2 | 160.967 g/mol
$174.25 - $667.29
Chemical Identifiers
| CAS | 91983-26-5 |
|---|---|
| Molecular Formula | C8H8BNO2 |
| Molecular Weight (g/mol) | 160.967 |
| MDL Number | MFCD01632200 |
| InChI Key | YKVMTTIYBSVPEQ-UHFFFAOYSA-N |
| Synonym | 4-cyanomethyl benzeneboronic acid, 4-cyanomethyl phenyl boronic acid, 4-cyanomethylphenylboronic acid, 4-cyanomethylphenyl boronic acid, 4-cyanomethyl phenylboronic acid, 4-boronophenylacetonitrile, boronic acid, 4-cyanomethyl phenyl, pubchem7789, acmc-209ree |
| PubChem CID | 2773342 |
| IUPAC Name | [4-(cyanomethyl)phenyl]boronic acid |
| SMILES | B(C1=CC=C(C=C1)CC#N)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1995503
|
Thermo Scientific Chemicals
L1995503 |
1 g |
Each for $174.25
|
|
|||||
|
AAL1995506
|
Thermo Scientific Chemicals
L1995506 |
5 g |
Each for $667.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 91983-26-5 | |
| 160.967 | |
| YKVMTTIYBSVPEQ-UHFFFAOYSA-N | |
| 2773342 | |
| B(C1=CC=C(C=C1)CC#N)(O)O |
| C8H8BNO2 | |
| MFCD01632200 | |
| 4-cyanomethyl benzeneboronic acid, 4-cyanomethyl phenyl boronic acid, 4-cyanomethylphenylboronic acid, 4-cyanomethylphenyl boronic acid, 4-cyanomethyl phenylboronic acid, 4-boronophenylacetonitrile, boronic acid, 4-cyanomethyl phenyl, pubchem7789, acmc-209ree | |
| [4-(cyanomethyl)phenyl]boronic acid |
Specifications
| 91983-26-5 | |
| C8H8BNO2 | |
| 1 g | |
| 4-cyanomethyl benzeneboronic acid, 4-cyanomethyl phenyl boronic acid, 4-cyanomethylphenylboronic acid, 4-cyanomethylphenyl boronic acid, 4-cyanomethyl phenylboronic acid, 4-boronophenylacetonitrile, boronic acid, 4-cyanomethyl phenyl, pubchem7789, acmc-209ree | |
| B(C1=CC=C(C=C1)CC#N)(O)O | |
| 160.967 | |
| 160.97 | |
| 4-(Cyanomethyl)benzeneboronic acid |
| ∼196°C to 200°C | |
| MFCD01632200 | |
| 3031323 | |
| YKVMTTIYBSVPEQ-UHFFFAOYSA-N | |
| [4-(cyanomethyl)phenyl]boronic acid | |
| 2773342 | |
| 98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only