missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6210303

 View more versions of this product

Catalog No. AAH6210303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester
1220219-61-3
2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl acetonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC#N)F
261.103
261.1
96%
1g
C14H17BFNO2
SAFMDYCIEJEGJQ-UHFFFAOYSA-N
2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
54759097
Solid

Chemical Identifiers

1220219-61-3
261.103
2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl acetonitrile
2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
C14H17BFNO2
SAFMDYCIEJEGJQ-UHFFFAOYSA-N
54759097
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC#N)F
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures

Documents