missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H62177MD

 View more versions of this product

Catalog No. AAH62177MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
4-Cyanomethyl-2-fluorobenzeneboronic acid pinacol ester used as a intermediate in pharmaceutical.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

4-Cyanomethyl-2-fluorobenzeneboronic acid pinacol ester
4-(Cyanomethyl)-2-fluorophenylboronic acid pinacol ester
Solid
250mg
261.1
96%

Chemical Identifiers

4-(Cyanomethyl)-2-fluorophenylboronic acid pinacol ester
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents