missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Cyanoindole, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442490010
Specifications
4-Cyanoindole | |
Glass Bottle | |
120°C to 121°C | |
16136-52-0 | |
100.0 | |
C9H6N2 | |
CEUFGDDOMXCXFW-UHFFFAOYSA-N | |
1H-indole-4-carbonitrile | |
3817602 | |
Powder |
Authentic | |
White to Brown | |
1g | |
96.0 | |
96% min. (HPLC) | |
1h-indol-4-yl cyanide, 4-cyano indole, 4-cyano-1h-indole, 4-cyanoindole, 4-indolecarbonitrile, acmc-209dmi, indole-4-carbonitrile, indole-4-cyano, pubchem7327 | |
C1=CC(=C2C=CNC2=C1)C#N | |
142.16 | |
142.16 | |
97% |
Chemical Identifiers
16136-52-0 | |
142.16 | |
1h-indol-4-yl cyanide, 4-cyano indole, 4-cyano-1h-indole, 4-cyanoindole, 4-indolecarbonitrile, acmc-209dmi, indole-4-carbonitrile, indole-4-cyano, pubchem7327 | |
1H-indole-4-carbonitrile |
C9H6N2 | |
CEUFGDDOMXCXFW-UHFFFAOYSA-N | |
3817602 | |
C1=CC(=C2C=CNC2=C1)C#N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger