Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanobutylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material 4-Cyanobutylzinc bromide
CAS 226570-68-9
Name Note Packaged under Argon in resealable ChemSeal™ bottles
Molecular Formula C5H8BrNZn
Linear Formula N≡C(CH2)4ZnBr
Formula Weight 227.41
MDL Number MFCD01316996
UN Number UN3399
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58897AD

  SDS
alfa aesar™
H58897AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.41
Quantity 50mL
Chemical Name or Material 4-Cyanobutylzinc bromide
Health Hazard 2 GHS H Statement
H225-H261-H314-H351-H302-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 226570-68-9
Concentration 0.5M in THF
UN Number UN3399
Molecular Formula C5H8BrNZn
Linear Formula N≡C(CH2)4ZnBr
MDL Number MFCD01316996
Recommended Storage 2° to 8°C
Health Hazard 1 Danger
Name Note Packaged under Argon in resealable ChemSeal™ bottles

Applications
It is used as primary and secondary intermediate.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and water.

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H351-H302-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.

2° to 8°C