missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Cyanobiphenyl, 95%, ACROS Organics™

Chemical Identifiers

CAS 2920-38-9
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C13H9N
Formula Weight 179.22
MDL Number MFCD00001821
Synonym 4-Phenylbenzonitrile
Chemical Name or Material 4-Cyanobiphenyl
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC346400050 SDS Thermo Fisher Scientific
346400050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2920-38-9
100.0
C13H9N
MFCD00001821
4-Cyanobiphenyl
94.0
95%
179.22
4-Phenylbenzonitrile
≥94%

Specifications

Crystalline Powder
5g
09, 672
100.0
C13H9N
95%
4-Phenylbenzonitrile
Authentic
≥94%
179.22
Beige
85° to 87°C
94.0
2920-38-9
MFCD00001821
Glass bottle
4-Cyanobiphenyl