Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Cyanobiphenyl-3-carboxylic acid, 95%

$89.14 - $358.65

Chemical Identifiers

Chemical Name or Material 4'-Cyanobiphenyl-3-carboxylic acid
CAS 149506-93-4
Molecular Formula C14H9NO2
Formula Weight 223.23
MDL Number MFCD04117352
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5190606

 
alfa aesar™
H5190606
5g Each for $358.65

AAH5190603

 
alfa aesar™
H5190603
1g Each for $89.14
Description & Specifications

Specifications

Formula Weight 223.23
Chemical Name or Material 4'-Cyanobiphenyl-3-carboxylic acid
Assay 95%
Molecular Formula C14H9NO2
MDL Number MFCD04117352
CAS 149506-93-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.