missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Cyanobenzyl alcohol, 97%

$25.24 - $329.47

Chemical Identifiers

CAS 874-89-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00870633
InChI Key XAASLEJRGFPHEV-UHFFFAOYSA-N
Synonym 4 hydroxymethyl-benzonitrile
PubChem CID 160549
IUPAC Name 4-(hydroxymethyl)benzonitrile
SMILES C1=CC(=CC=C1CO)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54283MD View Documents Alfa Aesar™
H54283MD
250mg Each for $25.24
 
AAH5428303 View Documents Alfa Aesar™
H5428303
1g Each for $79.08
 
AAH5428306 View Documents Alfa Aesar™
H5428306
5g Each for $329.47
 
Description

Description

Applications
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

874-89-5
133.15
XAASLEJRGFPHEV-UHFFFAOYSA-N
160549
C1=CC(=CC=C1CO)C#N
C8H7NO
MFCD00870633
4 hydroxymethyl-benzonitrile
4-(hydroxymethyl)benzonitrile

Specifications

32°C to 36°C
874-89-5
MFCD00870633
4 hydroxymethyl-benzonitrile
XAASLEJRGFPHEV-UHFFFAOYSA-N
4-(hydroxymethyl)benzonitrile
160549
97%
250mg
C8H7NO
UN3439
Insoluble in water.
C1=CC(=CC=C1CO)C#N
133.15
133.15
4-Cyanobenzyl alcohol
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures