Learn More
4-Cyanobenzyl alcohol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5428306
Description
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
4-Cyanobenzyl alcohol | |
32°C to 36°C | |
MFCD00870633 | |
UN3439 | |
Insoluble in water. | |
OCC1=CC=C(C=C1)C#N | |
133.15 | |
133.15 |
874-89-5 | |
C8H7NO | |
5 g | |
4-hydroxymethyl benzonitrile, 4-cyanobenzyl alcohol, p-cyanobenzyl alcohol, ccris 5116, benzonitrile, 4-hydroxymethyl, 4-cyanobenzenemethanol, 4-cyanophenyl methanol, 4-hydroxymethyl-benzonitrile, 4 hydroxymethyl-benzonitrile | |
XAASLEJRGFPHEV-UHFFFAOYSA-N | |
4-(hydroxymethyl)benzonitrile | |
160549 | |
97% |
Chemical Identifiers
874-89-5 | |
133.15 | |
XAASLEJRGFPHEV-UHFFFAOYSA-N | |
160549 | |
OCC1=CC=C(C=C1)C#N |
C8H7NO | |
MFCD00870633 | |
4-hydroxymethyl benzonitrile, 4-cyanobenzyl alcohol, p-cyanobenzyl alcohol, ccris 5116, benzonitrile, 4-hydroxymethyl, 4-cyanobenzenemethanol, 4-cyanophenyl methanol, 4-hydroxymethyl-benzonitrile, 4 hydroxymethyl-benzonitrile | |
4-(hydroxymethyl)benzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only